miniPCR® Lab Starter Pack Plus
Supplier: AMPLYUS LLC
QP-2500-09 QP-2516-26
470352-086EA 1944.79 USD
470352-086 470352-090
miniPCR® Lab Starter Pack Plus
Educational Classroom Kits and Activities
The essential tools of molecular biology in one handy bundle!
The miniPCR® Lab starter pack plus combines all the hardware you need to outfit your molecular biology lab with the reagents and consumables to get you up and running, enhanced with the inclusion of a Gyro™ Plus variable speed microcentrifuge.
Delivery information: The starter pack includes: miniPCR® mini8X or mini16X thermal cycler: a powerful, portable PCR machine always at your fingertips; blueGel™ system: all-in-one electrophoresis and transillumination; Gyro™ Plus: variable speed microcentrifuge; Set of 3 micropipettes (1 to 10 μl, 2 to 20 μl, 20 to 200 μl): Universal fit, compatible with standard micropipette tips, and easy to re-calibrate; Consumables starter pack: all the tubes, tips, and gel electrophoresis reagents you need to hit the ground running. Includes agarose (electrophoresis grade, 20 g), TBE electrophoresis buffer (powder to make 3 L), SeeGreen™ Nucleic Acid Gel Stain (20000X in water, 200 μl), microtubes (1.5 ml, bag of 500), PCR tubes (0.2 ml, bag of 100), 200 μl micropipette tips (2 racks of 96), and 10 μl micropipette tips (2 racks of 96).