2-Ethoxypyridin-4-amine 97%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-ethoxypyridine

A957266-250MG A957266-5G
76880-974EA 14 USD
76880-974 76880-976
2-Ethoxypyridin-4-amine 97%
2-Ethoxypyridin-4-amine

Caution: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD00235127
CAS Number: 89943-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR