2-Phenylbenzoxazole 98%

Supplier: Ambeed
Warning

A857803-1G A857803-250MG A857803-5G
76880-588EA 18 USD
76880-588 76880-590 76880-592
2-Phenylbenzoxazole 98%
2-Phenylbenzoxazole

Caution: for Research Use Only

Formula: C₁₃H₉NO
MDL Number: MFCD00012183
CAS Number: 833-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR