4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A604142-250MG A604142-100MG A604142-1G
76878-960EA 33 USD
76878-960 77884-453 77884-454
4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid 95%
4-Chlorothieno[3,2-d]pyrimidine-6-carboxylic acid

Caution: for Research Use Only

Formula: C₇H₃ClN₂O₂S
MDL Number: MFCD11846188
CAS Number: 875515-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR