(+)-JQ1 98%

Supplier: Ambeed
Warning

Synonyms: (S)-tert-Butyl 2-(4-(4-chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetate

A529842-100MG A529842-1MG A529842-250MG A529842-1G A529842-50MG A529842-5G
76878-296EA 65 USD
76878-296 76878-298 76878-300 77551-602 77673-410 77884-315
(+)-JQ1 98%
(+)-JQ1

Caution: for Research Use Only

Formula: C₂₃H₂₅ClN₄O₂S
MDL Number: MFCD22683748
CAS Number: 1268524-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR