D-Tagatose 98% + (mixture of isomers)

Supplier: Ambeed
Warning

Synonyms: (3S,4S,5R)-1,3,4,5,6-Pentahydroxyhexan-2-one

A453588-100G A453588-1G A453588-250MG A453588-25G A453588-5G
76877-540EA 168 USD
76877-540 76877-542 76877-544 76877-546 76877-548
D-Tagatose 98% + (mixture of isomers)
D-Tagatose

Caution: for Research Use Only

Formula: C₆H₁₂O₆
MDL Number: MFCD00134449
CAS Number: 87-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR