L(-)-Sorbose 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
L-(-)-Sorbose, (3S,4R,5S)-1,3,4,5,6-Pentahydroxyhexan-2-one
A385279-10G
A385279-25G
A385279-100G
76876-724EA
37
USD
76876-724
76876-726
77884-020
L(-)-Sorbose 95%
L(-)-Sorbose
Caution: for Research Use Only
Formula:
C₆H₁₂O₆ MW: 180.16 g/mol Boiling Pt: 380 |
MDL Number:
MFCD00151097 CAS Number: 87-79-6 Merck Index: 12,08874 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...