H-D-Leu-NH₂·HCl 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Leu-NH₂·HCl, H-D-Leu-NH2.HCl

A313215-1G A313215-5G A313215-25G
76875-782EA 24 USD
76875-782 76875-784 77612-806
H-D-Leu-NH₂·HCl 98%
H-D-Leu-NH₂·HCl

Caution: for Research Use Only

Formula: C₆H₁₅ClN₂O
MDL Number: MFCD00083640
CAS Number: 80970-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR