2-Hydroxymethylthiophen-4-boronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

A225250-100MG A225250-1G A225250-250MG A225250-5G
76874-072EA 40 USD
76874-072 76874-074 76874-076 77885-560
2-Hydroxymethylthiophen-4-boronic acid, pinacol ester 97%
2-Hydroxymethylthiophen-4-boronic acid, pinacol ester

Caution: for Research Use Only

Formula: C₁₁H₁₇BO₃S
MDL Number: MFCD08706011
CAS Number: 864754-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR