7-Hydroxycoumarin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Umbelliferone, 7-Hydroxy-2H-chromen-2-one, 7-Hydroxy-2-benzopyrone
A193753-25G
A193753-25MG
A193753-5G
76873-138EA
39.26
USD
76873-138
76873-140
76873-142
7-Hydroxycoumarin 98%
7-Hydroxycoumarin
Caution: for Research Use Only
Formula:
C₉H₆O₃ MW: 162.14 g/mol |
MDL Number:
MFCD00006878 CAS Number: 93-35-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...