5-Aminosalicylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Mesalamine, Mesalazine, 5-Amino-2-hydroxybenzoic acid, m-Aminosalicylic acid

A105320-100G A105320-10G A105320-25G A105320-500G
76869-394EA 18.04 USD
76869-394 76869-396 76869-398 76869-400
5-Aminosalicylic acid 98%
5-Aminosalicylic acid

Caution: for Research Use Only

Formula: C₇H₇NO₃
MW: 153.14 g/mol
MDL Number: MFCD00007877
CAS Number: 89-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR