4-Methoxythiophenol 97%

Supplier: Ambeed
Danger

A913850-25G A913850-10G A913850-100G A913850-500G
76845-664EA 41 USD
76845-664 77171-888 77893-715 77893-716
4-Methoxythiophenol 97%
4-Methoxythiophenol

Caution: for Research Use Only

Formula: C₇H₈OS
MDL Number: MFCD00004849
CAS Number: 696-63-9
UN: 3334
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR