4-(2-Chloroethyl)morpholine hydrochloride 95%

Supplier: Ambeed
Danger

Synonyms: N-(2-Chloroethyl)morpholine hydrochloride, N-(2-Chloroethyl)-Morpholinium Chloride

A443032-25G A443032-100G A443032-10G A443032-500G
76842-536EA 6 USD
76842-536 76979-390 76979-392 76979-394 76979-396
4-(2-Chloroethyl)morpholine hydrochloride 95%
4-(2-Chloroethyl)morpholine hydrochloride

Caution: For research use only

Formula: C₆H₁₃Cl₂NO
MW: 186.08 g/mol
MDL Number: MFCD00012797
CAS Number: 3647-69-6
UN: 2923
ADR: 8, 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR