Isoniazid 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Picolinic acid hydrazide, 4-Pyridinecarboxylic acid hydrazide, Isonicotinic hydrazide, Pyridine-4-carboxylic acid hydrazide, Isonicotinic acid hydrazide

A432881-25G A432881-100G A432881-10G A432881-500G A432881-1000G
76842-306EA 6 USD
76842-306 76979-314 76979-316 77594-342 77613-338
Isoniazid 98+%
Isoniazid

Caution: For research use only

Formula: C₆H₇N₃O
MW: 137.14 g/mol
MDL Number: MFCD00006426
CAS Number: 54-85-3
Merck Index: 13,05205

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR