1-Chloro-3-iodobenzene 98% stabilised

Supplier: Ambeed
Warning

Synonyms: 3-Chloroiodobenzene

Stabilization: Stabilised with copper chip

A289515-25G A289515-1000G A289515-100G A289515-10G A289515-1G A289515-500G A289515-5G
76837-556EA 13 USD
76837-556 77239-020 77239-022 77239-024 77239-026 77239-028 77239-030
1-Chloro-3-iodobenzene 98% stabilised
1-Chloro-3-iodobenzene

Caution: for Research Use Only

Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 230
Melting Pt: 54.5
MDL Number: MFCD00001046
CAS Number: 625-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR