(4-Fluoro-5-isopropyl-2-methoxyphenyl)boronic acid 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A250814-1G A250814-5G A250814-25G
76826-942EA 31.83 USD
76826-942 76826-944 77683-750
(4-Fluoro-5-isopropyl-2-methoxyphenyl)boronic acid 98%, Ambeed.Inc
(4-Fluoro-5-isopropyl-2-methoxyphenyl)boronic acid

Caution: For research use only.

Formula: C₁₀H₁₄BFO₃
MW: 212.02 g/mol
MDL Number: MFCD16294183
CAS Number: 875446-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR