2,6-Diethynylpyridine 98%

Supplier: Ambeed
Warning

A180417-1G A180417-250MG A180417-5G
76824-860EA 61 USD
76824-860 76824-862 76824-864
2,6-Diethynylpyridine 98%
2,6-Diethynylpyridine

Caution: for Research Use Only

Formula: C₉H₅N
MDL Number: MFCD09039127
CAS Number: 75867-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR