Alisertib 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4-((9-Chloro-7-(2-fluoro-6-methoxyphenyl)-5H-benzo[c]pyrimido[4,5-e]azepin-2-yl)amino)-2-methoxybenzoic acid, MLN8237

A102516-1MG A102516-250MG A102516-25MG A102516-10MG A102516-1G A102516-5MG A102516-100MG A102516-50MG
76821-284EA 12.74 USD
76821-284 76821-286 77570-138 77610-768 77610-770 77610-772 77682-942 77682-943
Alisertib 98%, Ambeed.Inc
Alisertib

Caution: For research use only.

Formula: C₂₇H₂₀ClFN₄O₄
MW: 518.92 g/mol
MDL Number: MFCD16621243
CAS Number: 1028486-01-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR