2,6-Dibromoanthracene 95%

Supplier: Ambeed
Warning

A631935-100MG A631935-1G A631935-250MG
76819-618EA 102.91 USD
76819-618 76819-620 76819-622
2,6-Dibromoanthracene 95%
2,6-Dibromoanthracene

Caution: for Research Use Only

Formula: C₁₄H₈Br₂
MW: 336.0213 g/mol
MDL Number: MFCD08276341
CAS Number: 186517-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR