Dimethoxy(diphenyl)silane 98%

Supplier: Ambeed
Warning

Synonyms: Dimethoxydiphenylsilane, Diphenyldimethoxysilane

A521449-100G A521449-25G A521449-500G A521449-1000G
76818-870EA 10 USD
76818-870 76818-872 76818-874 77884-303
Dimethoxy(diphenyl)silane 98%
Dimethoxy(diphenyl)silane

Caution: for Research Use Only

Formula: C₁₄H₁₆O₂Si
MDL Number: MFCD00025690
CAS Number: 6843-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR