2-Amino-4-bromo-6-chlorophenol 98%

Supplier: Ambeed
Warning

A387903-100MG A387903-1G A387903-250MG A387903-5G
76817-748EA 18.04 USD
76817-748 76817-750 76817-752 77672-926
2-Amino-4-bromo-6-chlorophenol 98%
2-Amino-4-bromo-6-chlorophenol

Caution: for Research Use Only

Formula: C₆H₅BrClNO
MW: 222.467 g/mol
MDL Number: MFCD16742724
CAS Number: 855836-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR