5-Bromo-N,N-diethylpicolinamide 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromopyridine-2-carboxylic acid diethylamide

A359975-1G A359975-250MG A359975-5G
76817-412EA 43 USD
76817-412 76817-414 76817-416
5-Bromo-N,N-diethylpicolinamide 95%
5-Bromo-N,N-diethylpicolinamide

Caution: for Research Use Only

Formula: C₁₀H₁₃BrN₂O
MW: 257.12 g/mol
MDL Number: MFCD07357420
CAS Number: 673485-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR