7-Methylguanine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-7-methyl-1H-purin-6(7H)-one, N7-Methylguanine

A295562-100MG A295562-250MG A295562-5G
76816-648EA 57.01 USD
76816-648 76816-650 77881-808
7-Methylguanine 97%
7-Methylguanine

Caution: for Research Use Only

Formula: C₆H₇N₅O
MDL Number: MFCD00038003
CAS Number: 578-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR