(±)-Mandelic acid 98%

Supplier: Ambeed
Warning

Synonyms: DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid

A228156-100G A228156-25G A228156-500G A228156-5G A228156-1000G
76815-094EA 18 USD
76815-094 76815-096 76815-098 76815-100 77672-020
(±)-Mandelic acid 98%
(±)-Mandelic acid

Caution: for Research Use Only

Formula: C₈H₈O₃
MDL Number: MFCD00064250
CAS Number: 90-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR