2-Amino-5-chlorobenzophenone 98%

Supplier: Ambeed
Warning

A153038-100G A153038-500G A153038-25G
76812-306EA 27 USD
76812-306 76812-308 77299-992
2-Amino-5-chlorobenzophenone 98%
2-Amino-5-chlorobenzophenone

Caution: for Research Use Only

Formula: C₁₃H₁₀ClNO
MDL Number: MFCD00007839
CAS Number: 719-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR