Phthalaldehyde 98%

Supplier: Ambeed
Danger

Synonyms: 1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde

A131637-10G A131637-25G A131637-100G
76811-218EA 6 USD
76811-218 76811-220 77611-164
Phthalaldehyde 98%
Phthalaldehyde

Caution: for Research Use Only

Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 266
Flash Pt: 132
MDL Number: MFCD00003335
CAS Number: 643-79-8
UN: 2923
ADR: 8,6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR