Isoeugenol 95%

Supplier: Ambeed
Warning

Synonyms: Propenylguaiacol, 4-Hydroxy-3-methoxy-1-propenylbenzene, 2-Methoxy-4-(prop-1-en-1-yl)phenol

A921294-100G A921294-10G A921294-25G
76808-020EA 25.47 USD
76808-020 76808-022 76845-674
Isoeugenol 95%
Isoeugenol

Caution: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00009285
CAS Number: 97-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR