Ferric pyrophosphate 82+%

Supplier: Ambeed
Warning

Synonyms: Iron(III) pyrophosphate, Iron (III) pyrophosphate

A886404-1000G A886404-100G A886404-25G A886404-500G
76807-888EA 78.51 USD
76807-888 76807-890 76807-892 76807-894
Ferric pyrophosphate 82+%
Ferric pyrophosphate

Caution: for Research Use Only

Formula: Fe₄O₂₁P₆
MW: 745.2100 g/mol
MDL Number: MFCD00016091
CAS Number: 10058-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR