Eriochrome Black T indicator

Supplier: Ambeed
Warning

Synonyms: 1-Naphthalenesulfonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), 1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate

A813732-1000G A813732-500G A813732-100G
76807-582EA 45.62 USD
76807-582 76807-584 77603-054
Eriochrome Black T indicator
Eriochrome Black T

Caution: for Research Use Only

Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
MDL Number: MFCD00003935
CAS Number: 1787-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR