(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
1-Fluoro-2,4-dinitrophenyl-5-L-alanine amide, N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, Propanamide, 2-[(5-fluoro-2,4-dinitrophenyl)amino]-, (2S)-
A774084-100MG
A774084-1G
A774084-250MG
A774084-5G
76807-332EA
11.67
USD
76807-332
76807-334
76807-336
76807-338
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide 97%
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide
Caution: for Research Use Only
Formula:
C₉H₉FN₄O₅ MW: 272.192 g/mol |
MDL Number:
MFCD00042049 CAS Number: 95713-52-3 UN: 1325 ADR: 4.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...