Ferroceneacetic acid 97%

Supplier: Ambeed
Warning

A719406-10G A719406-1G A719406-250MG A719406-5G
76806-832EA 315 USD
76806-832 76806-834 76806-836 76806-838
Ferroceneacetic acid 97%
Ferroceneacetic acid

Caution: for Research Use Only

Formula: C₁₂H₁₂FeO₂
MDL Number: MFCD00045464
CAS Number: 1287-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR