2-Amino-4-methylpentanamide hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-DL-Leu-NH₂·HCl, H-DL-Leu-NH2.HCl

A706844-250MG A706844-25G A706844-5G
76806-762EA 10 USD
76806-762 76806-764 76806-766
2-Amino-4-methylpentanamide hydrochloride 98%
2-Amino-4-methylpentanamide hydrochloride

Caution: for Research Use Only

Formula: C₆H₁₅ClN₂O
MDL Number: MFCD00070244
CAS Number: 10466-60-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR