Eugenol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole

A671082-1000G A671082-25G A671082-500G A671082-100G
76806-554EA 53 USD
76806-554 76806-556 76806-558 77573-160
Eugenol 98%
Eugenol

Caution: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 255
Melting Pt: -10
Flash Pt: 110
MDL Number: MFCD00008654
CAS Number: 97-53-0
Merck Index: 13,03931

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR