Famciclovir 97%

Supplier: Ambeed
Warning

Synonyms: 2-[2-(2-Amino-9H-purin-9-yl)ethyl]-1,3-propanediol Diacetate, 2-(2-(2-Amino-9H-purin-9-yl)ethyl)propane-1,3-diyl diacetate

A643497-100MG A643497-1G A643497-50MG A643497-5G
76806-332EA 6.37 USD
76806-332 76806-334 76806-336 76806-338
Famciclovir 97%
Famciclovir

Caution: for Research Use Only

Formula: C₁₄H₁₉N₅O₄
MW: 321.34 g/mol
MDL Number: MFCD00866964
CAS Number: 104227-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR