Ethyl cinnamate 99%

Supplier: Ambeed
Warning

Synonyms: NSC 6773, 3-Phenyl-acrylicacidethylester, Ethyl 3-phenyl-2-propenoate, Cinnamic acid ethyl ester, 3-Phenyl-2-propenoic acid ethyl ester

A631671-1000G A631671-100G A631671-500G A631671-25G
76806-220EA 59 USD
76806-220 76806-222 76806-224 77308-010
Ethyl cinnamate 99%
Ethyl cinnamate

Caution: for Research Use Only

Formula: C₁₁H₁₂O₂
MW: 176.21 g/mol
Boiling Pt: 271
Melting Pt: 7
MDL Number: MFCD00009189
CAS Number: 103-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR