Dimethyldimyristylammonium bromide 98%
Supplier: Ambeed
|
Warning
|
A584431-100G
A584431-1G
A584431-25G
A584431-5G
76805-834EA
127
USD
76805-834
76805-836
76805-838
76805-840
Dimethyldimyristylammonium bromide 98%
Dimethyldimyristylammonium bromide
Caution: for Research Use Only
Formula:
C₃₀H₆₄BrN |
MDL Number:
MFCD00191429 CAS Number: 68105-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...