di-Sodium L(+)-tartrate dihydrate 99%

Supplier: Ambeed
Warning

Synonyms: Sodium L-(+)-tartrate dihydrate, L(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Tartaric acid disodium salt dihydrate, (R,R)-(+)-Sodium tartrate dibasic dihydrate, Disodium L-(+)-Tartrate Dihydrate, (R,R)-(+)-Disodium tartrate dihydrate, L(+)-Tartaric acid, disodium salt, dihydrate, Sodium L-tartrate dibasic dihydrate, L(+)-Sodium tartrate dibasic dihydrate, Sodium Tartrate, Dihydrate, (+)-Disodium L-tartrate dihydrate, (+)-Disodium-L-tartrat dihydrate

A519649-25G A519649-5G
76805-152EA 10.61 USD
76805-152 76805-154
di-Sodium L(+)-tartrate dihydrate 99%
di-Sodium L(+)-tartrate dihydrate

Caution: for Research Use Only

Formula: NaOOC(CHOH)₂COONa·2H₂O
MW: 230.08 g/mol
MDL Number: MFCD00150035
CAS Number: 6106-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR