Indole 98%

Supplier: Ambeed
Danger

Synonyms: 1H-Indole

A493773-25G A493773-100G
76804-932EA 11 USD
76804-932 77602-132
Indole 98%
Indole

Caution: for Research Use Only

Formula: C₈H₇N
MW: 117.15 g/mol
Flash Pt: 110
MDL Number: MFCD00005607
CAS Number: 120-72-9
UN: 2811
ADR: 6.1 Ⅲ
Merck Index: 13,04985

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR