7-Amino-4-(trifluoromethyl)coumarin 99%

Supplier: Ambeed
Warning

A490087-1G A490087-25G A490087-5G
76804-866EA 15 USD
76804-866 76804-868 76804-870
7-Amino-4-(trifluoromethyl)coumarin 99%
7-Amino-4-(trifluoromethyl)coumarin

Caution: for Research Use Only

Formula: C₁₀H₆F₃NO₂
MDL Number: MFCD00006858
CAS Number: 53518-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR