Diphenyliodonium hexafluorophosphate 98%
Supplier: Ambeed
|
Danger
|
A433077-1G
A433077-25G
A433077-5G
76804-300EA
7
USD
76804-300
76804-302
76804-304
Diphenyliodonium hexafluorophosphate 98%
Diphenyliodonium hexafluorophosphate
Caution: for Research Use Only
Formula:
C₁₂H₁₀F₆IP |
MDL Number:
MFCD00061398 CAS Number: 58109-40-3 UN: 2923 ADR: 8,6.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...