Girard's reagent P (carbazoylmethylpyridinium chloride) 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-(2-Hydrazinyl-2-oxoethyl)pyridin-1-ium chloride, Girard's Reagent P, 1-(Hydrazinocarbonylmethyl)pyridinium chloride, (1-Pyridinio)acetohydrazide chloride
A425840-10G
A425840-1G
A425840-25G
A425840-5G
76804-192EA
61.54
USD
76804-192
76804-194
76804-196
76804-198
Girard's reagent P (carbazoylmethylpyridinium chloride) 95%
Girard's reagent P (Carbazoylmethylpyridinium chloride)
Caution: for Research Use Only
Formula:
C₇H₁₀ClN₃O MW: 187.63 g/mol |
MDL Number:
MFCD00011992 CAS Number: 1126-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...