Methoxytrimethylsilane 98%

Supplier: Ambeed
Danger

A407382-25G A407382-5G
76804-000EA 23 USD
76804-000 76804-002
Methoxytrimethylsilane 98%
Methoxytrimethylsilane

Caution: for Research Use Only

Formula: C₄H₁₂OSi
MDL Number: MFCD00008344
CAS Number: 1825-61-2
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR