Eugenol acetate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Allyl-2-methoxyphenyl acetate, Acetyleugenol

A387785-100G A387785-25G A387785-25MG A387785-5MG A387785-500G
76803-804EA 49.87 USD
76803-804 76803-806 76803-808 76803-810 77668-796
Eugenol acetate 98%
Eugenol acetate

Caution: for Research Use Only

Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD00026191
CAS Number: 93-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR