Linalyl acetate 98%

Supplier: Ambeed
Warning

A378791-100G A378791-25G A378791-500G
76803-698EA 15 USD
76803-698 76803-700 77879-821
Linalyl acetate 98%
Linalyl acetate

Caution: for Research Use Only

Formula: C₁₂H₂₀O₂
MDL Number: MFCD00008907
CAS Number: 115-95-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR