Lithium carbonate 99%
Supplier: Ambeed
|
Warning
|
A281259-1000G
A281259-100G
A281259-25G
A281259-500G
A281259-50MG
A281259-5G
76802-184EA
136
USD
76802-184
76802-186
76802-188
76802-190
76802-192
76802-194
Lithium carbonate 99%
Lithium carbonate
Caution: for Research Use Only
Formula:
Li₂CO₃ MW: 73.89 g/mol Melting Pt: 720 |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 Merck Index: 13,05549 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...