1-Propyl-4-ethynylbenzene 97%

Supplier: Ambeed
Warning

Synonyms: 1-Ethynyl-4-propylbenzene, (4-Propylphenyl)acetylene, 4-n-Propylphenylacetylene, (4-Propylphenyl)ethyne

A209453-100G A209453-10G A209453-1G A209453-25G A209453-5G
76800-588EA 103 USD
76800-588 76800-590 76800-592 76800-594 76800-596
1-Propyl-4-ethynylbenzene 97%
1-Propyl-4-ethynylbenzene

Caution: for Research Use Only

Formula: C₁₁H₁₂
MDL Number: MFCD00173886
CAS Number: 62452-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR