Diethyl sebacate 95%

Supplier: Ambeed
Warning

A205509-100G A205509-250MG A205509-25G A205509-500G A205509-1000G
76800-500EA 9 USD
76800-500 76800-502 77626-040 77626-042 77879-280
Diethyl sebacate 95%
Diethyl sebacate

Caution: for Research Use Only

Formula: C₁₄H₂₆O₄
MDL Number: MFCD00009219
CAS Number: 110-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR