Ethyldiphenylphosphine 98%

Supplier: Ambeed
Warning

A162325-100G A162325-1G A162325-25G A162325-5G
76799-050EA 173 USD
76799-050 76799-052 76799-054 76799-056
Ethyldiphenylphosphine 98%
Ethyldiphenylphosphine

Caution: for Research Use Only

Formula: C₁₄H₁₅P
MDL Number: MFCD00015170
CAS Number: 607-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR