2,5-Piperazinedione 95%

Supplier: Ambeed
Warning

A146595-100G A146595-25G A146595-500G A146595-5G
76798-432EA 92 USD
76798-432 76798-434 76798-436 76798-438
2,5-Piperazinedione 95%
2,5-Piperazinedione

Caution: for Research Use Only

Formula: C₄H₆N₂O₂
MDL Number: MFCD00006009
CAS Number: 106-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR