Ebselen 98%

Supplier: Ambeed
Danger

Synonyms: 2-Phenyl-1,2-benzoisoselenazol-3(2H)-one, 2-Phenylbenzo[d][1,2]selenazol-3(2H)-one

A121080-100MG A121080-1G A121080-1MG A121080-250MG A121080-50MG A121080-5MG A121080-10MG
76797-354EA 73 USD
76797-354 76797-356 76797-358 76797-360 76797-362 76797-364 77599-746
Ebselen 98%
Ebselen

Caution: for Research Use Only

Formula: C₁₃H₉NOSe
MW: 274.20 g/mol
MDL Number: MFCD00210937
CAS Number: 60940-34-3
UN: 3283
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR